CH6371378515 Chem Impex International, Inc.

Boc-O-benzyl-D-tyrosine-N-hydroxysuccinimide ester

  • MDL number MFCD01631982
  • Molecular Weight : 468.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-155 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-tyrosine-i-n-i-hydroxysuccinimide-ester/2740?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-d-tyrosine-i-n-i-hydroxysuccinimide-ester/2740?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart