CH6371378520 Chem Impex International, Inc.

Boc-L-aspartic acid a-amide

  • MDL number MFCD00076946
  • Cas Number : 74244-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-144° C
Purity 98.5
Storage temp. Store at 0-8°C
Physical Form White crystal powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-a-amide/2755?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-a-amide/2755?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart