CH6371378527 Chem Impex International, Inc.

Boc-L-aspartic acid beta-cyclopentyl ester

  • MDL number MFCD00057818
  • Cas Number : 71447-58-0
  • Molecular Weight : 301.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-92 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-beta-cyclopentyl-ester/2762?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-aspartic-acid-beta-cyclopentyl-ester/2762?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart