CH6371378532 Chem Impex International, Inc.

Fmoc-L-a-aminoadipic acid

  • MDL number MFCD01631983

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-135
Synonym Fmoc-L-Aad-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-alpha-aminoadipic-acid/2768?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-alpha-aminoadipic-acid/2768?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart