CH6371378533 Chem Impex International, Inc.

Fmoc-D-a-aminoadipic acid

  • MDL number MFCD01317727
  • Cas Number : 218457-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-147° C
Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-a-aminoadipic-acid/2769?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-a-aminoadipic-acid/2769?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart