CH6371378534 Chem Impex International, Inc.

Boc-D-a-aminoadipic acid

  • MDL number MFCD00797554
  • Cas Number : 110544-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Aad-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-a-aminoadipic-acid/2770?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-a-aminoadipic-acid/2770?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart