CH6371378539 Chem Impex International, Inc.

3-Benzothienyl-L-alanine

  • MDL number MFCD00079684
  • Cas Number : 72120-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-L-Ala(3-benzothienyl)-OH
Purity 98.5
Storage temp. Store at 0-8
Physical Form Almost white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-benzothienyl-l-alanine/2777?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-benzothienyl-l-alanine/2777?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart