CH6371378540 Chem Impex International, Inc.

3-Benzothienyl-D-alanine

  • MDL number MFCD00079683
  • Cas Number : 111139-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-240 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form Almost white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-benzothienyl-d-alanine/2778?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-benzothienyl-d-alanine/2778?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart