CH6371378545 Chem Impex International, Inc.

Boc-p-phenyl-D-Phenylalanine

  • MDL number MFCD00191185
  • Cas Number : 128779-47-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-125� C
Synonym Boc-D-Bip(4,4')-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-p-i-phenyl-d-phenylalanine/2783?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-p-i-phenyl-d-phenylalanine/2783?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart