CH6371378547 Chem Impex International, Inc.

Fmoc-p-phenyl-D-Phenylalanine

  • MDL number MFCD00191199
  • Cas Number : 205526-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Bip(4,4')-OH
Purity 0.995
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-p-i-phenyl-d-phenylalanine/2785?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-p-i-phenyl-d-phenylalanine/2785?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart