CH6371378553 Chem Impex International, Inc.

Fmoc-4-bromo-D-phenylalanine

  • MDL number MFCD00273459
  • Cas Number : 198545-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-177
Synonym Fmoc-D-Phe(4-Br)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-bromo-d-phenylalanine/2791?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-bromo-d-phenylalanine/2791?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart