CH6371378555 Chem Impex International, Inc.

2-Chloro-D-phenylalanine

  • MDL number MFCD00077920
  • Cas Number : 80126-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-215� C
Synonym D-Phe(2-Cl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-chloro-d-phenylalanine/2793?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-chloro-d-phenylalanine/2793?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart