CH6371378564 Chem Impex International, Inc.

Fmoc-3-chloro-L-phenylalanine

  • MDL number MFCD00672548
  • Cas Number : 198560-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-112
Synonym Fmoc-L-Phe(3-Cl)-OH
Purity 99.7
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-chloro-l-phenylalanine/2802?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-chloro-l-phenylalanine/2802?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart