CH6371378565 Chem Impex International, Inc.

Fmoc-3-chloro-D-phenylalanine

  • MDL number MFCD00672549
  • Cas Number : 205526-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121 - 126 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-chloro-d-phenylalanine/2803?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-chloro-d-phenylalanine/2803?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart