CH6371378577 Chem Impex International, Inc.

Fmoc-4-cyano-D-phenylalanine

  • MDL number MFCD00672560
  • Cas Number : 205526-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-189
Synonym Fmoc-4-cyano-D-phenylalanine
Purity 98
Storage temp. Store at 0-8
Physical Form White product
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-cyano-d-phenylalanine/2815?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-cyano-d-phenylalanine/2815?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart