CH6371378590 Chem Impex International, Inc.

Fmoc-3,5-difluoro-D-phenylalanine

  • MDL number MFCD00797580
  • Cas Number : 205526-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Phe(3,5-DiF)-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-5-difluoro-d-phenylalanine/2828?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-5-difluoro-d-phenylalanine/2828?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart