CH6371378596 Chem Impex International, Inc.

Fmoc-3,3-diphenyl-D-alanine

  • MDL number MFCD00672338
  • Cas Number : 189937-46-0
  • Molecular Weight : 463.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-3-diphenyl-d-alanine/2834?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-3-diphenyl-d-alanine/2834?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart