CH6371378597 Chem Impex International, Inc.

2-Fluoro-D-phenylalanine

  • MDL number MFCD00066447
  • Cas Number : 122839-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-200� C
Synonym D-Phe(2-F)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/2-fluoro-d-phenylalanine/2835?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/2-fluoro-d-phenylalanine/2835?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart