CH6371378605 Chem Impex International, Inc.

Fmoc-3-fluoro-D-phenylalanine

  • MDL number MFCD00672554
  • Cas Number : 198545-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-160
Synonym Fmoc-D-Phe(3-F)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-fluoro-d-phenylalanine/2843?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-fluoro-d-phenylalanine/2843?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart