CH6371378608 Chem Impex International, Inc.

Fmoc-D-beta-indanylglycine

  • MDL number MFCD00797587
  • Cas Number : 205526-40-5
  • Molecular Weight : 413.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-189 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-ß-indanylglycine/2847?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-ß-indanylglycine/2847?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart