CH6371378622 Chem Impex International, Inc.

Fmoc-O-2-chlorotrityl-L-tyrosine

  • MDL number MFCD01073759
  • Cas Number : 350241-80-4
  • Molecular Weight : 680.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 160-165
Purity 99
Storage temp. Store at 0-8
Physical Form White to yellow powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-2-chlorotrityl-l-tyrosine/2958?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-2-chlorotrityl-l-tyrosine/2958?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart