CH6371378624 Chem Impex International, Inc.

4-Methyltrityl chloride

  • MDL number MFCD01073758
  • Cas Number : 23429-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-101
Synonym 4-Methyltriphenylmethyl chloride
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white crystals
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-methyltrityl-chloride/2960?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-methyltrityl-chloride/2960?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart