CH6371378629 Chem Impex International, Inc.

Acetyl-L-alanine methyl amide

  • MDL number MFCD00038238
  • Cas Number : 19701-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-183 �C
Synonym Ac-L-Ala-NHMe
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-alanine-methyl-amide/2968?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-alanine-methyl-amide/2968?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart