CH6371378630 Chem Impex International, Inc.

Acetyl-4-amino-L-phenylalanine methyl ester hydrochloride

  • MDL number MFCD19707041
  • Cas Number : 36097-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-106 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Pale pink powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-4-amino-l-phenylalanine-methyl-ester-hydrochloride/2970?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-4-amino-l-phenylalanine-methyl-ester-hydrochloride/2970?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart