CH6371378634 Chem Impex International, Inc.

Nalpha-Acetyl-Ngamma-trityl-L-asparagine

  • MDL number MFCD00236745
  • Cas Number : 163277-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-217 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-acetyl-i-n-i-sup-sup-trityl-l-asparagine/2975?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-acetyl-i-n-i-sup-sup-trityl-l-asparagine/2975?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart