CH6371378637 Chem Impex International, Inc.

Acetyl-4-bromo-DL-phenylalanine

  • MDL number MFCD00237505
  • Cas Number : 273730-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-189� C
Synonym Ac-DL-Phe(4-Br)-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-4-bromo-dl-phenylalanine/2980?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-4-bromo-dl-phenylalanine/2980?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart