CH6371378642 Chem Impex International, Inc.

Na-Ac-Nd-trityl-L-glutamine

  • MDL number MFCD00236753
  • Cas Number : 163277-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-214� C
Synonym Ac-L-Gln(Trt)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-ac-i-n-i-sup-d-sup-trityl-l-glutamine/2987?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-ac-i-n-i-sup-d-sup-trityl-l-glutamine/2987?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart