CH6371378644 Chem Impex International, Inc.

Acetyl-L-glutamic acid g-tert-butyl ester

  • MDL number MFCD00236754
  • Cas Number : 84192-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-L-Glu(OtBu)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-glutamic-acid-gamma-i-tert-i-butyl-ester/2991?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-glutamic-acid-gamma-i-tert-i-butyl-ester/2991?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart