CH6371378647 Chem Impex International, Inc.

Na-Acetyl-Nim-trityl-L-histidine

  • MDL number MFCD00236758
  • Cas Number : 183498-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-L-His(Trt)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-acetyl-i-n-i-sup-im-sup-trityl-l-histidine/2995?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-acetyl-i-n-i-sup-im-sup-trityl-l-histidine/2995?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart