CH6371378648 Chem Impex International, Inc.

Acetyl-L-isoleucine amide

  • MDL number MFCD00080832
  • Cas Number : 56711-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-isoleucine-amide/2996?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-isoleucine-amide/2996?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart