CH6371378649 Chem Impex International, Inc.

Acetyl-4-iodo-D-phenylalanine

  • MDL number MFCD00236759
  • Cas Number : 201351-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-204
Synonym Ac-D-Phe(4-I)-OH
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-4-iodo-d-phenylalanine/2998?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-4-iodo-d-phenylalanine/2998?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart