CH6371378656 Chem Impex International, Inc.

Acetyl-L-methionine sulfoxide

  • MDL number MFCD00236766
  • Cas Number : 108646-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-132
Synonym Ac-L-Met(O)-OH
Purity 95
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-methionine-sulfoxide/3011?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-methionine-sulfoxide/3011?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart