CH6371378657 Chem Impex International, Inc.

Acetyl-O-methyl-L-tyrosine methyl ester

  • MDL number MFCD00069961
  • Cas Number : 17355-24-7
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103-109 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-i-o-i-methyl-l-tyrosine-methyl-ester/3012?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-i-o-i-methyl-l-tyrosine-methyl-ester/3012?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart