CH6371378660 Chem Impex International, Inc.

Acetyl-D-phenylalanine methyl ester

  • MDL number MFCD00026172
  • Cas Number : 21156-62-7
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-92
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/acetyl-d-phenylalanine-methyl-ester/3016?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-d-phenylalanine-methyl-ester/3016?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart