CH6371378661 Chem Impex International, Inc.

Acetyl-DL-phenylglycine

  • MDL number MFCD00055924
  • Cas Number : 15962-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-DL-Phg-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-dl-phenylglycine/3019?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-dl-phenylglycine/3019?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart