CH6371378664 Chem Impex International, Inc.

Acetyl-L-serine methyl ester

  • MDL number MFCD00038508
  • Cas Number : 54322-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-L-Ser-OMe
Purity 0.98
Storage temp. Store at 0-8
Physical Form Light yellow syrup
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-serine-methyl-ester/3023?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-serine-methyl-ester/3023?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart