CH6371378665 Chem Impex International, Inc.

Acetyl-O-tert-butyl-L-threonine

  • MDL number MFCD00236771
  • Cas Number : 163277-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-141� C
Synonym Ac-L-Thr(tBu)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-i-o-i-i-tert-i-butyl-l-threonine/3025?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-i-o-i-i-tert-i-butyl-l-threonine/3025?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart