CH6371378669 Chem Impex International, Inc.

Formyl-L-alanine

  • MDL number MFCD00037357
  • Cas Number : 10512-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-140 �C
Synonym For-L-Ala-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/formyl-l-alanine/3031?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/formyl-l-alanine/3031?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart