CH6371378670 Chem Impex International, Inc.

N-Methyl-D-alanine hydrochloride

  • MDL number MFCD00797880

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-Me-D-Ala-OH�
Purity 98
Storage temp. Store at 0-8
Physical Form White to off white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-d-alanine-hydrochloride/3032?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-d-alanine-hydrochloride/3032?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart