CH6371378677 Chem Impex International, Inc.

Ngamma-Trityl-L-asparagine hydrate

  • MDL number MFCD00153422
  • Cas Number : 132388-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-212 °C
Purity 97
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-g-sup-trityl-l-asparagine-hydrate/3039?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-g-sup-trityl-l-asparagine-hydrate/3039?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart