CH6371378678 Chem Impex International, Inc.

L-Aspartic acid a-tert-butyl ester hydrochloride

  • MDL number MFCD00153424
  • Cas Number : 176164-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Asp-OtBu HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-a-i-tert-i-butyl-ester-hydrochloride/3042?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-a-i-tert-i-butyl-ester-hydrochloride/3042?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart