CH6371378679 Chem Impex International, Inc.

L-Aspartic acid diethyl ester hydrochloride

  • MDL number MFCD00038910
  • Cas Number : 16115-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-111
Synonym L-Asp(OEt)-OEt HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-aspartic-acid-diethyl-ester-hydrochloride/3043?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-aspartic-acid-diethyl-ester-hydrochloride/3043?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart