CH6371378687 Chem Impex International, Inc.

Nd-Trityl-D-glutamine hydrate

  • MDL number MFCD00153433
  • Cas Number : 200625-76-9
  • Molecular Weight : 406.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 218-222° C
Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-d-sup-trityl-d-glutamine-hydrate/3051?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-d-sup-trityl-d-glutamine-hydrate/3051?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart