CH6371378688 Chem Impex International, Inc.

Glycine tert-butyl ester acetate salt

  • MDL number MFCD00153441
  • Cas Number : 38024-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52 - 54 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/glycine-i-tert-i-butyl-ester-acetate-salt/3052?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/glycine-i-tert-i-butyl-ester-acetate-salt/3052?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart