CH6371378689 Chem Impex International, Inc.

Nalpha,Nim-Ditrityl-L-histidine

  • MDL number MFCD00153443
  • Cas Number : 74853-62-6
  • Molecular Weight : 639.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183 - 203 °C
Purity 99
Storage temp. Store at = -4 °C
Physical Form Off-white to yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-n-i-sup-im-sup-ditrityl-l-histidine/3053?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-n-i-sup-im-sup-ditrityl-l-histidine/3053?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart