CH6371378691 Chem Impex International, Inc.

L-Isoleucine allyl ester 4-toluenesulfonate salt

  • MDL number MFCD00043317
  • Cas Number : 88224-05-9
  • Molecular Weight : 343.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118 - 122 °C
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-isoleucine-allyl-ester-4-toluenesulfonate-salt/3055?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-isoleucine-allyl-ester-4-toluenesulfonate-salt/3055?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart