CH6371378693 Chem Impex International, Inc.

L-Lysine amide dihydrochloride

  • MDL number MFCD00058287
  • Cas Number : 51127-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Lys-NH2 2HCl
Purity 0.95
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-lysine-amide-dihydrochloride/3057?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-lysine-amide-dihydrochloride/3057?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart