CH6371378694 Chem Impex International, Inc.

L-Phenylalanine allyl ester 4-toluenesulfonate salt

  • MDL number MFCD00043306
  • Cas Number : 88224-00-4
  • Molecular Weight : 377.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-160 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-phenylalanine-allyl-ester-4-toluenesulfonate-salt/3058?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-phenylalanine-allyl-ester-4-toluenesulfonate-salt/3058?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart