CH6371378705 Chem Impex International, Inc.

Nin-Boc-D-tryptophan

  • MDL number MFCD00237295
  • Cas Number : 201290-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Trp(Boc)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Light pale fine powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-in-sup-boc-d-tryptophan/3070?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-in-sup-boc-d-tryptophan/3070?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart