CH6371378719 Chem Impex International, Inc.

Ala-Gly-OMe HCl

  • MDL number MFCD00155457
  • Cas Number : 23404-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 983
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-gly-ome·hcl/3086?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-gly-ome·hcl/3086?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart